Anti-SQSTM1 Mouse Monoclonal Antibody [clone: OTI4G9]
Proveedor: OriGene
ORIGTA502132EA 603 EUR
ORIGTA502132
Anti-SQSTM1 Mouse Monoclonal Antibody [clone: OTI4G9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: SQSTM1 (sequestosome 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now