Anti-ANXA3 Mouse Monoclonal Antibody [clone: OTI2F3]

Proveedor: OriGene
ORIGTA502122EA 581 EUR
ORIGTA502122
Anti-ANXA3 Mouse Monoclonal Antibody [clone: OTI2F3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: ANXA3 (annexin A3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR