Anti-GRHPR Mouse Monoclonal Antibody [clone: OTI9E6]

Proveedor: OriGene
ORIGTA502088EA 603 EUR
ORIGTA502088
Anti-GRHPR Mouse Monoclonal Antibody [clone: OTI9E6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500; Flow Cytometry: 1:100

Type: Primary
Antigen: GRHPR (glyoxylate reductase/hydroxypyruvate reductase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR