Anti-GRHPR Mouse Monoclonal Antibody [clone: OTI1A9]

Proveedor: OriGene
ORIGTA502086EA 608 EUR
ORIGTA502086
Anti-GRHPR Mouse Monoclonal Antibody [clone: OTI1A9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: GRHPR (glyoxylate reductase/hydroxypyruvate reductase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR