Anti-DUPD1 Mouse Monoclonal Antibody [clone: OTI7E4]

Proveedor: OriGene
ORIGTA502080EA 608 EUR
ORIGTA502080
Anti-DUPD1 Mouse Monoclonal Antibody [clone: OTI7E4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: DUPD1 (dual specificity phosphatase and pro isomerase domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR