Anti-RASD2 Mouse Monoclonal Antibody [clone: OTI1F7]

Proveedor: OriGene
ORIGTA502072EA 603 EUR
ORIGTA502072
Anti-RASD2 Mouse Monoclonal Antibody [clone: OTI1F7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RASD2 (RASD family, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR