Anti-SDR9C7 Mouse Monoclonal Antibody [clone: OTI10F11]

Proveedor: OriGene
ORIGTA502015EA 603 EUR
ORIGTA502015
Anti-SDR9C7 Mouse Monoclonal Antibody [clone: OTI10F11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SDR9C7 (short chain dehydrogenase/reductase family 9C, member 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR