Anti-KEAP1 Mouse Monoclonal Antibody [clone: OTI1G2]

Proveedor: OriGene
TA501989
ORIGTA501989EA 616 EUR
ORIGTA501989
Anti-KEAP1 Mouse Monoclonal Antibody [clone: OTI1G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KEAP1 (kelch-like ECH-associated protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR