Anti-FAM84B Mouse Monoclonal Antibody [clone: OTI5G1]

Proveedor: OriGene
ORIGTA501969EA 603 EUR
ORIGTA501969
Anti-FAM84B Mouse Monoclonal Antibody [clone: OTI5G1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FAM84B (family with sequence similarity 84, member B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR