Anti-HAO1 Mouse Monoclonal Antibody [clone: OTI5F4]

Proveedor: OriGene
ORIGTA501957EA 603 EUR
ORIGTA501957
Anti-HAO1 Mouse Monoclonal Antibody [clone: OTI5F4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: HAO1 (hydroxyacid oxidase (glycolate oxidase) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR