Anti-SULT1A1 Mouse Monoclonal Antibody [clone: OTI2C5]

Proveedor: OriGene
ORIGTA501952EA 603 EUR
ORIGTA501952
Anti-SULT1A1 Mouse Monoclonal Antibody [clone: OTI2C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT1A1 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR