Anti-OTUB2 Mouse Monoclonal Antibody [clone: OTI11C8]

Proveedor: OriGene
TA501945
ORIGTA501945EA 616 EUR
ORIGTA501945
Anti-OTUB2 Mouse Monoclonal Antibody [clone: OTI11C8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: OTUB2 (OTU deubiquitinase, ubiquitin aldehyde binding 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR