Anti-OTUB2 Mouse Monoclonal Antibody [clone: OTI6F7]
Proveedor: OriGene
ORIGTA501942EA 603 EUR
ORIGTA501942
Anti-OTUB2 Mouse Monoclonal Antibody [clone: OTI6F7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: OTUB2 (OTU deubiquitinase, ubiquitin aldehyde binding 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human