Anti-OSBPL11 Mouse Monoclonal Antibody [clone: OTI5A10]

Proveedor: OriGene
ORIGTA501940EA 603 EUR
ORIGTA501940
Anti-OSBPL11 Mouse Monoclonal Antibody [clone: OTI5A10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: OSBPL11 (oxysterol binding protein-like 11)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR