Anti-HAO1 Mouse Monoclonal Antibody [clone: OTI3E7]

Proveedor: OriGene
ORIGTA501937EA 616 EUR
ORIGTA501937
Anti-HAO1 Mouse Monoclonal Antibody [clone: OTI3E7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:2000 Flow Cytometry: 1:50-1:100

Type: Primary
Antigen: HAO1 (hydroxyacid oxidase (glycolate oxidase) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR