Anti-FAM84B Mouse Monoclonal Antibody [clone: OTI5A9]

Proveedor: OriGene
ORIGTA501918EA 603 EUR
ORIGTA501918
Anti-FAM84B Mouse Monoclonal Antibody [clone: OTI5A9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: FAM84B (family with sequence similarity 84, member B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR