Anti-BIN3 Mouse Monoclonal Antibody [clone: OTI7D6]

Proveedor: OriGene
ORIGTA501910EA 581 EUR
ORIGTA501910
Anti-BIN3 Mouse Monoclonal Antibody [clone: OTI7D6]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BIN3 (bridging integrator 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR