Anti-SLFNL1 Mouse Monoclonal Antibody [clone: OTI3D4]

Proveedor: OriGene
ORIGTA501884EA 603 EUR
ORIGTA501884
Anti-SLFNL1 Mouse Monoclonal Antibody [clone: OTI3D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SLFNL1 (schlafen-like 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR