Anti-DUPD1 Mouse Monoclonal Antibody [clone: OTI1F3]
Proveedor: OriGene
ORIGTA501857EA 603 EUR
ORIGTA501857
Anti-DUPD1 Mouse Monoclonal Antibody [clone: OTI1F3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: DUPD1 (dual specificity phosphatase and pro isomerase domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human