Anti-MICAL1 Mouse Monoclonal Antibody [clone: OTI6H10]

Proveedor: OriGene
ORIGTA501845EA 603 EUR
ORIGTA501845
Anti-MICAL1 Mouse Monoclonal Antibody [clone: OTI6H10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100; Immunochemistry: 1:150

Type: Primary
Antigen: MICAL1 (microtubule associated monooxygenase, calponin and LIM domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR