Anti-PBLD Mouse Monoclonal Antibody [clone: OTI4A5]

Proveedor: OriGene
ORIGTA501840EA 603 EUR
ORIGTA501840
Anti-PBLD Mouse Monoclonal Antibody [clone: OTI4A5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PBLD (phenazine biosynthesis-like protein domain containing)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR