Anti-GSTT2 Mouse Monoclonal Antibody [clone: OTI3B6]

Proveedor: OriGene
ORIGTA501832EA 581 EUR
ORIGTA501832
Anti-GSTT2 Mouse Monoclonal Antibody [clone: OTI3B6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GSTT2 (glutathione S-transferase theta 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR