Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI3G5]

Proveedor: OriGene
ORIGTA501828EA 603 EUR
ORIGTA501828
Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI3G5]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR