Anti-BEST3 Mouse Monoclonal Antibody [clone: OTI1B6]

Proveedor: OriGene
ORIGTA501826EA 581 EUR
ORIGTA501826
Anti-BEST3 Mouse Monoclonal Antibody [clone: OTI1B6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BEST3 (bestrophin 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR