Anti-GAS7 Mouse Monoclonal Antibody [clone: OTI6G5]

Proveedor: OriGene
ORIGTA501825EA 581 EUR
ORIGTA501825
Anti-GAS7 Mouse Monoclonal Antibody [clone: OTI6G5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GAS7 (growth arrest-specific 7)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR