Anti-TCP11L2 Mouse Monoclonal Antibody [clone: OTI1H7]

Proveedor: OriGene
ORIGTA501822EA 581 EUR
ORIGTA501822
Anti-TCP11L2 Mouse Monoclonal Antibody [clone: OTI1H7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: TCP11L2 (t-complex 11, testis-specific-like 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR