Anti-AFP Mouse Monoclonal Antibody [clone: OTI2A6]

Proveedor: OriGene
ORIGTA501784EA 603 EUR
ORIGTA501784
Anti-AFP Mouse Monoclonal Antibody [clone: OTI2A6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-500

Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR