Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI5H2]
Proveedor: OriGene
ORIGTA501763EA 593 EUR
ORIGTA501763
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI5H2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PIK3AP1 (phosphoinositide-3-kinase adaptor protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human