Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI5D4]

Proveedor: OriGene
ORIGTA501753EA 581 EUR
ORIGTA501753
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI5D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: PIK3AP1 (phosphoinositide-3-kinase adaptor protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR