Anti-UHMK1 Mouse Monoclonal Antibody [clone: OTI2D3]

Proveedor: OriGene
ORIGTA501719EA 603 EUR
ORIGTA501719
Anti-UHMK1 Mouse Monoclonal Antibody [clone: OTI2D3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: UHMK1 (U2AF homology motif (UHM) kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR