Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI3D4]

Proveedor: OriGene
ORIGTA501714EA 616 EUR
ORIGTA501714
Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI3D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: SULT2A1 (sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR