Anti-RIT2 Mouse Monoclonal Antibody [clone: OTI4A6]

Proveedor: OriGene
ORIGTA501701EA 603 EUR
ORIGTA501701
Anti-RIT2 Mouse Monoclonal Antibody [clone: OTI4A6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RIT2 (Ras-like without CAAX 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR