Anti-RND1 Mouse Monoclonal Antibody [clone: OTI1C3]

Proveedor: OriGene
ORIGTA501691EA 608 EUR
ORIGTA501691
Anti-RND1 Mouse Monoclonal Antibody [clone: OTI1C3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RND1 (Rho family GTPase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR