Anti-HP Mouse Monoclonal Antibody [clone: OTI2B11]

Proveedor: OriGene
ORIGTA501675EA 603 EUR
ORIGTA501675
Anti-HP Mouse Monoclonal Antibody [clone: OTI2B11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: HP (haptoglobin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR