Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI2A6]

Proveedor: OriGene
ORIGTA501658EA 603 EUR
ORIGTA501658
Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI2A6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT2A1 (sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR