Anti-AK5 Mouse Monoclonal Antibody [clone: OTI2A6]

Proveedor: OriGene
ORIGTA501650EA 581 EUR
ORIGTA501650
Anti-AK5 Mouse Monoclonal Antibody [clone: OTI2A6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: AK5 (adenylate kinase 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR