Anti-UBE2T Mouse Monoclonal Antibody [clone: OTI2F5]

Proveedor: OriGene
ORIGTA501645EA 608 EUR
ORIGTA501645
Anti-UBE2T Mouse Monoclonal Antibody [clone: OTI2F5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: UBE2T (ubiquitin-conjugating enzyme E2T (putative))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR