Anti-EPHX2 Mouse Monoclonal Antibody [clone: OTI1H5]

Proveedor: OriGene
ORIGTA501627EA 603 EUR
ORIGTA501627
Anti-EPHX2 Mouse Monoclonal Antibody [clone: OTI1H5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: EPHX2 (epoxide hydrolase 2, cytoplasmic)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR