Anti-CBWD1 Mouse Monoclonal Antibody [clone: OTI9B8]

Proveedor: OriGene
ORIGTA501621EA 581 EUR
ORIGTA501621
Anti-CBWD1 Mouse Monoclonal Antibody [clone: OTI9B8]
Anticuerpos
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CBWD1 (COBW domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR