Anti-NT5DC1 Mouse Monoclonal Antibody [clone: OTI2H8]

Proveedor: OriGene
ORIGTA501586EA 603 EUR
ORIGTA501586
Anti-NT5DC1 Mouse Monoclonal Antibody [clone: OTI2H8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NT5DC1 (5'-nucleotidase domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR