Anti-VWA5A Mouse Monoclonal Antibody [clone: OTI3E6]

Proveedor: OriGene
ORIGTA501570EA 603 EUR
ORIGTA501570
Anti-VWA5A Mouse Monoclonal Antibody [clone: OTI3E6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: VWA5A (von Willebrand factor A domain containing 5A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR