Anti-NT5DC1 Mouse Monoclonal Antibody [clone: OTI3E1]

Proveedor: OriGene
ORIGTA501567EA 603 EUR
ORIGTA501567
Anti-NT5DC1 Mouse Monoclonal Antibody [clone: OTI3E1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NT5DC1 (5'-nucleotidase domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR