Anti-C16orf72 Mouse Monoclonal Antibody [clone: OTI1A2]

Proveedor: OriGene
ORIGTA501562EA 593 EUR
ORIGTA501562
Anti-C16orf72 Mouse Monoclonal Antibody [clone: OTI1A2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: C16orf72 (chromosome 16 open reading frame 72)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR