Anti-TUBA3E Mouse Monoclonal Antibody [clone: OTI2A8]

Proveedor: OriGene
ORIGTA501545EA 603 EUR
ORIGTA501545
Anti-TUBA3E Mouse Monoclonal Antibody [clone: OTI2A8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TUBA3E (tubulin, alpha 3e)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR