Anti-QPRT Mouse Monoclonal Antibody [clone: OTI4E5]

Proveedor: OriGene
TA501519
ORIGTA501519EA 616 EUR
ORIGTA501519
Anti-QPRT Mouse Monoclonal Antibody [clone: OTI4E5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: QPRT (quinolinate phosphoribosyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR