Anti-LIN7B Mouse Monoclonal Antibody [clone: OTI1C9]

Proveedor: OriGene
TA501516
ORIGTA501516EA 662 EUR
ORIGTA501516
Anti-LIN7B Mouse Monoclonal Antibody [clone: OTI1C9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LIN7B (lin-7 homolog B (C. elegans))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR