Anti-LIN7B Mouse Monoclonal Antibody [clone: OTI1G2]

Proveedor: OriGene
ORIGTA501513EA 603 EUR
ORIGTA501513
Anti-LIN7B Mouse Monoclonal Antibody [clone: OTI1G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: LIN7B (lin-7 homolog B (C. elegans))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR