Anti-CRYM Mouse Monoclonal Antibody [clone: OTI1B5]

Proveedor: OriGene
ORIGTA501433EA 603 EUR
ORIGTA501433
Anti-CRYM Mouse Monoclonal Antibody [clone: OTI1B5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:300; Immunofluorescence: 1:100

Type: Primary
Antigen: CRYM (crystallin, mu)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR