Anti-MIOX Mouse Monoclonal Antibody [clone: OTI1A7]

Proveedor: OriGene
ORIGTA501420EA 581 EUR
ORIGTA501420
Anti-MIOX Mouse Monoclonal Antibody [clone: OTI1A7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: MIOX (myo-inositol oxygenase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR