Anti-PKLR Mouse Monoclonal Antibody [clone: OTI1G9]

Proveedor: OriGene
ORIGTA501418EA 616 EUR
ORIGTA501418
Anti-PKLR Mouse Monoclonal Antibody [clone: OTI1G9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PKLR (pyruvate kinase, liver and RBC)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR